Alkali Scientific ELISA Plates Stripwells, White Frame with Clear Wells, Flat Bottom, 5/pk, 50/cs
Numéro de catalogue. 502336232
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials and they conform to SBS/ ANSI standards. The plates arrive with low evaporation lids and are perfectly suited for multichanneling. They have a maximum well volume of 325ul. They offer excellent well uniformity, CV <5%.
Numéro de catalogue.
502336232
Fournisseur
Alkali ScientificCode fournisseur
MP26
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
ELISA Plates Stripwells are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials and they conform to SBS/ ANSI standards. The plates arrive with low evaporation lids and are perfectly suited for multichanneling. They have a maximum well volume of 325ul. They offer excellent well uniformity, CV <5%.