Learn More
Corning™ 96-well Clear Flat Bottom CellBIND™-treated Polystyrene Sterile Microplate with Lid
Numéro de catalogue. 01001030
Change view
Click to view available options
Sterility:
Sterile
Numéro de catalogue. Sterility
01001030 Sterile
1 options
Numéro de catalogue. 01001030 Fournisseur Corning™ Code fournisseur 3809
Il en reste null
This microplate features a bottom that is 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Bottom is 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well cross-talk
  • Optically clear flat bottom permits direct microscopic viewing
Spécifications
Barcode No Barcode
Format Half-Area
Lid Yes
Array 8 x 12
Color White, Clear Bottom
Product Type Microplate
Material Plastic
Volume (Metric) Well 360 μL
No. of Wells 96
Sterility Sterile
Surface Treatment CellBIND
Well Shape Flat Bottom (F-well)
No. per Case 100
No. per Pack 20
Well Color Clear
Afficher plus de résultats Afficher moins de résultats