Learn More
Fisherbrand™ 95kPA Biohazard Mailers
For shipments of biological samples by mail and common carriers
$817.17 - $1,741.14
Spécifications
| Certifications/Compliance | Exempt Human Specimen, Exempt Animal Specimen and Biological Substance |
|---|---|
| Applications | For transport of biological substances |
Avertissement:
These products do not meet the packaging requirements for Biological Substance Category A materials and do not contain the absorbent materials, cushioning materials, and markings required to ship Biological Substances Category B materials.
| Numéro de catalogue | Type | Prix | Quantity & Availability | ||||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Type | Prix | Quantity & Availability | ||||||
|
22130542
|
Single Tube 95kPa Mailer |
caisse de 24 chaque for $817.17
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||||
|
22130541
|
Multiple Tube 95kPa Mailer |
caisse de 50 chaque for $1,741.14
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||||
Description
Fisherbrand 95kPa Biohazard Dual Mailers are designed for specimen mailing, common carrier shipping and transport within local, regional and national courier networks.
- For shipments of biological samples by mail and common carriers
- Accommodates a variety of blood tubes and other routine laboratory containers
Small Single Tube Dual Mailer (Catalog No. 22-130-542)
- Accommodates a range of standard blood collection tubes from 13 × 75 to 16 × 100mm
- Inner receptacle dimensions: 0.75 × 5 in. (I.D. × Length)
- Outer fiberboard mailer dimensions: 1.75 × 7.75 in. (I.D. × Length)
- Two standard blood collection tubes ranging in size from 13 × 75 to 16 × 100mm
- 15 and 50mL centrifuge tubes, slide specimens and fecal transport vials
- Inner receptacle dimensions: 2.5 × 4.25 in. (I.D. × Length)
- Outer Fiberboard mailer dimensions: 2 × 4.75 in. (I.D. × Length)
- Will accept up to four standard blood collection tubes ranging in size from 13 × 75mm to 16 × 100mm
- 15 and 50mL centrifuge tubes
- Slide specimens, fecal transport vials and many other confirmatory test sample containers
- Inner receptacle dimensions: 2.125 × 4.75 in. (I.D. × Length)
- Outer fiberboard mailer dimensions: 3.25 × 7 in. (I.D. × Length)
Compliance
Exempt Human Specimen, Exempt Animal Specimen and Biological Substance
Spécifications
| Exempt Human Specimen, Exempt Animal Specimen and Biological Substance | |
| For transport of biological substances |