missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-9H-carbazole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. P23515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23515G 5 g
P23511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23515G Supplier TCI America Supplier No. P23515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87220-68-6
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.319
InChI Key ITPJDBJBKAFUEG-UHFFFAOYSA-N
Synonym 3-Formyl-9-phenyl-9H-carbazole
PubChem CID 18515440
IUPAC Name 9-phenylcarbazole-3-carbaldehyde
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C=O)C4=CC=CC=C42

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
Formula Weight 271.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-9H-carbazole-3-carboxaldehyde