missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(Hydroxymethyl)anthracene 98.0+%, TCI America™
Supplier: TCI America H0670250G
Specifications
| 9-(Hydroxymethyl)anthracene | |
| 163°C | |
| C15H12O | |
| 250 g | |
| JCJNNHDZTLRSGN-UHFFFAOYSA-N | |
| anthracen-9-ylmethanol | |
| 73848 | |
| ≥98.0% (GC) |
| 1468-95-7 | |
| Yellow | |
| MFCD00001264 | |
| 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO | |
| 208.26 | |
| 208.26 | |
| Crystalline Powder |
Chemical Identifiers
| 1468-95-7 | |
| 208.26 | |
| JCJNNHDZTLRSGN-UHFFFAOYSA-N | |
| 73848 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO |
| C15H12O | |
| MFCD00001264 | |
| 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297 | |
| anthracen-9-ylmethanol |
Safety and Handling
RTECSNumber : CB0577500
TSCA : Yes