missing translation for 'onlineSavingsMsg'
Learn More

9-(Hydroxymethyl)anthracene 98.0+%, TCI America™

Catalog No. p-7130663
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
H067025G 25 g
H0670250G 250 g
2 options

Catalog No. H067025G

Supplier: TCI America H067025G

Only null left

Chemical Identifiers

CAS 1468-95-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00001264
InChI Key JCJNNHDZTLRSGN-UHFFFAOYSA-N
Synonym 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
PubChem CID 73848
IUPAC Name anthracen-9-ylmethanol
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

Specifications

Melting Point 163°C
Color Yellow
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(Hydroxymethyl)anthracene
RTECSNumber CB0577500
TSCA Yes