missing translation for 'onlineSavingsMsg'
Learn More

9-Heptadecanone 98,0+%, TCI America™

Numéro de catalogue. H05365G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H05365G 5 g
H053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H05365G Fournisseur TCI America Code fournisseur H05365G

Identifiants chimiques

CAS 540-08-9
Molecular Formula C17H34O
Molecular Weight (g/mol) 254.458
MDL Number MFCD00009573
InChI Key WTJKUFMLQFLJOT-UHFFFAOYSA-N
Synonym 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2
PubChem CID 10887
IUPAC Name heptadecan-9-one
SMILES CCCCCCCCC(=O)CCCCCCCC

Spécifications

Melting Point 54°C
Color Blanc-jaune
Quantity 5 g
Formula Weight 254.46
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 9-Heptadecanone
EINECSNumber (2)-2475
RTECSNumber MI3925000
TSCA No