missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl Pentafluorophenyl Carbonate 98.0+%, TCI America™

Catalog No. F09365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09365G 5 g
F09361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09365G Supplier TCI America Supplier No. F09365G

Chemical Identifiers

CAS 88744-04-1
Molecular Formula C21H11F5O3
Molecular Weight (g/mol) 406.308
MDL Number MFCD00013261
InChI Key CBBKZVZOEBSFQX-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl pentafluorophenyl carbonate, fmoc-opfp, 9h-fluoren-9-yl methyl perfluorophenyl carbonate, 9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl carbonate, 9h-fluoren-9-ylmethyl pentafluorophenyl carbonate, carbonic acid 9h-fluoren-9-ylmethyl pentafluorophenyl ester, carbonic acid,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl ester, acmc-20ecck, fmoc pentafluorophenyl ester, 9-fluorenylmethylpentafluorophenylcarbonate
PubChem CID 2733201
IUPAC Name 9H-fluoren-9-ylmethyl (2,3,4,5,6-pentafluorophenyl) carbonate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)OC4=C(C(=C(C(=C4F)F)F)F)F

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 406.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethyl Pentafluorophenyl Carbonate