missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. F08045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08045G 5 g
F08041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08045G Supplier TCI America Supplier No. F08045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6223-83-2
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.215
MDL Number MFCD00001145
InChI Key AFQYQSWTVCNJQT-UHFFFAOYSA-N
Synonym 9-fluorenone-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid, 9h-fluorene-4-carboxylic acid, 9-oxo, 9-fluorenone-4-carboxlic acid, 9-oxofluorene-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid #
PubChem CID 80361
IUPAC Name 9-oxofluorene-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=CC=C3C(=O)O

Specifications

Melting Point 228°C
Color Yellow
Quantity 5 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone-4-carboxylic Acid