missing translation for 'onlineSavingsMsg'
Learn More

9-Bromophenanthrene, 96%

Catalog No. AC107241000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107241000 100 g Glass bottle
AC107240100 10 g Glass bottle
AC107240250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107241000 Supplier Thermo Scientific Chemicals Supplier No. 107241000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 573-17-1 | C14H9Br | 257.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 573-17-1
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001174
InChI Key RSQXKVWKJVUZDG-UHFFFAOYSA-N
Synonym 9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene
PubChem CID 11309
IUPAC Name 9-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Yellow
Boiling Point 180°C to 190°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 05, 671
Formula Weight 257.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 9-Bromophenanthrene, 96%
Show More Show Less
EINECSNumber 209-351-6
RTECSNumber SF7197000

RUO – Research Use Only