missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(Bromomethyl)nonadecane 96.0+%, TCI America™
Supplier: TCI America B518825ML
Specifications
| 9-(Bromomethyl)nonadecane | |
| 195°C | |
| MFCD28515435 | |
| XSQSDBVMLJNZKU-UHFFFAOYSA-N | |
| 9-(bromomethyl)nonadecane | |
| 23148745 | |
| ≥96.0% (GC) |
| 69620-20-8 | |
| C20H41Br | |
| 25 mL | |
| CCCCCCCCCCC(CCCCCCCC)CBr | |
| 361.452 | |
| 361.45 | |
| Liquid |
Chemical Identifiers
| 69620-20-8 | |
| 361.452 | |
| XSQSDBVMLJNZKU-UHFFFAOYSA-N | |
| 9-(bromomethyl)nonadecane |
| C20H41Br | |
| MFCD28515435 | |
| 23148745 | |
| CCCCCCCCCCC(CCCCCCCC)CBr |
Safety and Handling
TSCA : No