accessibility menu, dialog, popup

UserName

H3CCH3Br

CAS RN 69620-20-8

IUPAC Name: 9-(bromomethyl)nonadecane
Synonyms: 9-(bromomethyl)nonadecane
Molecular Weight (g/mol): 361.45
Molecular Formula: C20H41Br
InChi Key: XSQSDBVMLJNZKU-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(CBr)CCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results