missing translation for 'onlineSavingsMsg'
Learn More

9-(Bromomethyl)nonadecane 96.0+%, TCI America™

Catalog No. p-7124005
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51885ML 5 mL
B518825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51885ML Supplier TCI America Supplier No. B51885ML
Only null left

Chemical Identifiers

CAS 69620-20-8
Molecular Formula C20H41Br
Molecular Weight (g/mol) 361.452
MDL Number MFCD28515435
InChI Key XSQSDBVMLJNZKU-UHFFFAOYSA-N
PubChem CID 23148745
IUPAC Name 9-(bromomethyl)nonadecane
SMILES CCCCCCCCCCC(CCCCCCCC)CBr

Specifications

Boiling Point 195°C
Quantity 5 mL
Formula Weight 361.45
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-(Bromomethyl)nonadecane