missing translation for 'onlineSavingsMsg'
Learn More

9-Bromofluorene 98.0+%, TCI America™

Catalog No. B10465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10465G 5 g
B104625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10465G Supplier TCI America Supplier No. B10465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1940-57-4
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001133
InChI Key AHCDKANCCBEQJJ-UHFFFAOYSA-N
Synonym 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene
PubChem CID 16024
IUPAC Name 9-bromo-9H-fluorene
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br

Specifications

Melting Point 104°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 245.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromofluorene
RTECSNumber LL5890000
TSCA No