missing translation for 'onlineSavingsMsg'
Learn More

9-Bromofluorene, 97+%

Catalog No. AAA1042818
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1042818 50 g
AAA1042806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1042818 Supplier Thermo Scientific Chemicals Supplier No. A1042818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1940-57-4 | C13H9Br | 245.119 g/mol

9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers

Chemical Identifiers

CAS 1940-57-4
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001133
InChI Key AHCDKANCCBEQJJ-UHFFFAOYSA-N
Synonym 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene
PubChem CID 16024
IUPAC Name 9-bromo-9H-fluorene
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br

Specifications

Melting Point 101°C to 105°C
Odor Irritating
Quantity 50 g
UN Number UN3261
Beilstein 2047220
Solubility Information Insoluble in water.
Formula Weight 245.12
Percent Purity ≥98%
Chemical Name or Material 9-Bromofluorene
Hazard Category H314-H317
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 217-722-9
RTECSNumber LL5890000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only