missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromo-9-phenylfluorene 96.0+%, TCI America™
Supplier: TCI America B170225G
Specifications
| 9-Bromo-9-phenylfluorene | |
| 100°C | |
| C19H13Br | |
| 25 g | |
| 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl | |
| C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br | |
| 321.217 | |
| 321.22 | |
| Crystalline Powder |
| 55135-66-5 | |
| Yellow | |
| MFCD00075522 | |
| 3261 | |
| HYQXNCDBSALQLB-UHFFFAOYSA-N | |
| 9-bromo-9-phenylfluorene | |
| 231624 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 55135-66-5 | |
| 321.217 | |
| HYQXNCDBSALQLB-UHFFFAOYSA-N | |
| 231624 | |
| C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br |
| C19H13Br | |
| MFCD00075522 | |
| 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl | |
| 9-bromo-9-phenylfluorene |
Safety and Handling
TSCA : No