missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene 96.0+%, TCI America™

Catalog No. p-7119213
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17025G 5 g
B170225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17025G Supplier TCI America Supplier No. B17025G
Only null left

Chemical Identifiers

CAS 55135-66-5
Molecular Formula C19H13Br
Molecular Weight (g/mol) 321.217
MDL Number MFCD00075522
InChI Key HYQXNCDBSALQLB-UHFFFAOYSA-N
Synonym 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
PubChem CID 231624
IUPAC Name 9-bromo-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br

Specifications

Melting Point 100°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 321.22
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-9-phenylfluorene