accessibility menu, dialog, popup

UserName

Br

CAS RN 55135-66-5

IUPAC Name: 9-bromo-9-phenyl-9H-fluorene
Synonyms: 9-bromo-9-phenylfluorene
Molecular Weight (g/mol): 321.22
Molecular Formula: C19H13Br
InChi Key: HYQXNCDBSALQLB-UHFFFAOYSA-N
SMILES: BrC1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)