missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine Hydrate 98.0+%, TCI America™

Catalog No. A273950MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A273950MG 50 mg
A273910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A273950MG Supplier TCI America Supplier No. A273950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38819-10-2
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00065486
InChI Key NYHBQMYGNKIUIF-FJFJXFQQSA-N
Synonym Ara-G, Guanine 9-beta-D-Arabinofuranoside
PubChem CID 65412
ChEBI CHEBI:73141
IUPAC Name 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 225°C
Color White
Quantity 50 mg
Formula Weight 283.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-beta-D-Arabinofuranosylguanine Hydrate
RTECSNumber MF8301000
TSCA No
Recommended Storage Refrigerator