missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenemethanol, 97%

Catalog No. AAA1518830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1518830 250 g
AAA1518809 10 g
AAA1518818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1518830 Supplier Thermo Scientific Chemicals Supplier No. A1518830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1468-95-7 | C15H12O | 208.26 g/mol

An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 1468-95-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00001264
InChI Key JCJNNHDZTLRSGN-UHFFFAOYSA-N
Synonym 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
PubChem CID 73848
IUPAC Name anthracen-9-ylmethanol
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

Specifications

Melting Point 160°C to 165°C
Quantity 250 g
Beilstein 1873402
Solubility Information Soluble in hot methanol very faint turbidity.
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 9-Anthracenemethanol
EINECSNumber 215-998-5
RTECSNumber CB0577500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only