missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Aminofluorene Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A062225G
Specifications
| 9-Aminofluorene Hydrochloride | |
| 257°C | |
| C13H12ClN | |
| 25 g | |
| SYKJOJSYQSVNOM-UHFFFAOYSA-N | |
| hydrogen 9H-fluoren-9-amine chloride | |
| 2724685 | |
| ≥98.0% (HPLC,T) |
| 5978-75-6 | |
| White-Yellow | |
| MFCD00012536 | |
| 9h-fluoren-9-amine hydrochloride, 9-aminofluorene hydrochloride, 9h-fluoren-9-amine, hydrochloride, 9-fluorenamine hydrochloride, 9-aminofluorene hcl, fluorene-9-ylamine, chloride, 9h-fluoren-9-amine hcl, acmc-1b10k, 9-aminoflu-orene hydrochloride, 9-aminofluorene monohydrochloride | |
| [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 217.70 | |
| 217.70 | |
| Crystalline Powder |
Chemical Identifiers
| 5978-75-6 | |
| 217.70 | |
| SYKJOJSYQSVNOM-UHFFFAOYSA-N | |
| 2724685 | |
| [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12 |
| C13H12ClN | |
| MFCD00012536 | |
| 9h-fluoren-9-amine hydrochloride, 9-aminofluorene hydrochloride, 9h-fluoren-9-amine, hydrochloride, 9-fluorenamine hydrochloride, 9-aminofluorene hcl, fluorene-9-ylamine, chloride, 9h-fluoren-9-amine hcl, acmc-1b10k, 9-aminoflu-orene hydrochloride, 9-aminofluorene monohydrochloride | |
| hydrogen 9H-fluoren-9-amine chloride |
Safety and Handling
TSCA : No