accessibility menu, dialog, popup

UserName

HClNH2

CAS RN 5978-75-6

IUPAC Name: hydrogen 9H-fluoren-9-amine chloride
Synonyms: hydrogen 9H-fluoren-9-amine chloride
Molecular Weight (g/mol): 217.7
Molecular Formula: C13H12ClN
InChi Key: SYKJOJSYQSVNOM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)