missing translation for 'onlineSavingsMsg'
Learn More

9-Aminofluorene Hydrochloride 98.0+%, TCI America™

Catalog No. p-7116970
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06225G 5 g
A062225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06225G Supplier TCI America Supplier No. A06225G

Chemical Identifiers

CAS 5978-75-6
Molecular Formula C13H12ClN
Molecular Weight (g/mol) 217.70
MDL Number MFCD00012536
InChI Key SYKJOJSYQSVNOM-UHFFFAOYSA-N
Synonym 9h-fluoren-9-amine hydrochloride, 9-aminofluorene hydrochloride, 9h-fluoren-9-amine, hydrochloride, 9-fluorenamine hydrochloride, 9-aminofluorene hcl, fluorene-9-ylamine, chloride, 9h-fluoren-9-amine hcl, acmc-1b10k, 9-aminoflu-orene hydrochloride, 9-aminofluorene monohydrochloride
PubChem CID 2724685
IUPAC Name hydrogen 9H-fluoren-9-amine chloride
SMILES [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 257°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Aminofluorene Hydrochloride