missing translation for 'onlineSavingsMsg'
Learn More

9-Aminocamptothecin 95.0+%, TCI America™

Catalog No. A206310MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A206310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A206310MG Supplier TCI America Supplier No. A206310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91421-43-1
Molecular Formula C20H17N3O4
Molecular Weight (g/mol) 363.37
MDL Number MFCD00909855
InChI Key FUXVKZWTXQUGMW-FQEVSTJZSA-N
PubChem CID 72402
ChEBI CHEBI:80755
IUPAC Name (19S)-8-amino-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=C(N)C1=C3)C2=O

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 363.37
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Aminocamptothecin
RTECSNumber UQ0490500
TSCA No