missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Dimethylfluorene 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 97°C |
| Color | White |
| Boiling Point | 287°C |
| Quantity | 1 g |
| Formula Weight | 194.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,9-Dimethylfluorene |
Sécurité et manipulation
| TSCA | No |