missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)-10-(1-naphthyl)anthracene 98.0+%, TCI America™

Catalog No. p-7123909
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5029200MG 200 mg
B50291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5029200MG Supplier TCI America Supplier No. B5029200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1160506-32-0
Molecular Formula C30H19Br
Molecular Weight (g/mol) 459.386
MDL Number MFCD26400490
InChI Key DMAFDYCUZMVPNZ-UHFFFAOYSA-N
PubChem CID 88951361
IUPAC Name 9-(4-bromophenyl)-10-naphthalen-1-ylanthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC=C(C=C6)Br

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 459.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Bromophenyl)-10-(1-naphthyl)anthracene