missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(3-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 110°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 398.30 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9-(3-Biphenylyl)-3-bromocarbazole |
Sécurité et manipulation
| TSCA | No |