missing translation for 'onlineSavingsMsg'
Learn More

9-(2-Biphényl)-10-bromoanthracène 98,0+%, TCI America™

Numéro de catalogue. p-7123823
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B48951G 1 g
B48955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B48951G Fournisseur TCI America Code fournisseur B48951G

Identifiants chimiques

CAS 400607-16-1
Molecular Formula C26H17Br
Molecular Weight (g/mol) 409.33
MDL Number MFCD28138083
InChI Key NANUBXRTTQXXDS-UHFFFAOYSA-N
PubChem CID 22138280
IUPAC Name 9-{[1,1'-biphényl]-2-yl}-10-bromoanthracène
SMILES BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC=CC=C1C1=CC=CC=C1

Spécifications

Melting Point 148°C
Quantity 1 g
Formula Weight 409.33
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 9-(2-Biphenylyl)-10-bromoanthracene