missing translation for 'onlineSavingsMsg'
Learn More

9,10-Diphénylanthracine 98,0+%, TCI America™

Numéro de catalogue. D16891G
Modifier l'affichage
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D16891G 1 g
D168925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D16891G Fournisseur TCI America Code fournisseur D16891G

Identifiants chimiques

CAS 1499-10-1
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD00001253
InChI Key FCNCGHJSNVOIKE-UHFFFAOYSA-N
Synonym anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard
PubChem CID 15159
ChEBI CHEBI:51676
IUPAC Name 9,10-diphénylanthracène
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Spécifications

Color Jaune
Boiling Point 485°C
Quantity 1 g
Formula Weight 330.43
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 9,10-Diphenylanthracene
EINECSNumber (4)-1873
RTECSNumber CB0545500
TSCA Yes