missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydro-9,9-dimethylacridine 98.0+%, TCI America™

Catalog No. D50285G
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50285G 5 g
D5028200MG 200 mg
D50281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D50285G Supplier TCI America Supplier No. D50285G
Only null left

Chemical Identifiers

CAS 6267-02-3
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
MDL Number MFCD00030130
InChI Key JSEQNGYLWKBMJI-UHFFFAOYSA-N
Synonym 9,9-Dimethyl-9,10-dihydroacridine
PubChem CID 22647
IUPAC Name 9,9-dimethyl-10H-acridine
SMILES CC1(C2=CC=CC=C2NC3=CC=CC=C31)C

Specifications

Melting Point 127°C
Color White-Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 209.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydro-9,9-dimethylacridine