missing translation for 'onlineSavingsMsg'
Learn More

9,10-Di(1-naphthayl)anthracene (purified by sublimation) 99.0+%, TCI America™

Catalog No. D50651G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50651G Supplier TCI America Supplier No. D50651G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26979-27-1
Molecular Formula C34H22
Molecular Weight (g/mol) 430.55
MDL Number MFCD04222155
InChI Key GWNJZSGBZMLRBW-UHFFFAOYSA-N
PubChem CID 259845
IUPAC Name 9,10-dinaphthalen-1-ylanthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC=CC7=CC=CC=C76

Specifications

Melting Point 360°C
Color White-Yellow
Quantity 1 g
Formula Weight 430.55
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Di(1-naphthayl)anthracene (purified by sublimation)