Learn More
Agilent BioTek 800 TS Absorbance Reader
GREENER_CHOICE
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
Supplier:  Agilent BioTek 800TSNBSN
 Afficher plus de versions de ce produit
Numéro de catalogue. BT800TSNB
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Warranty
12 months
Spécifications
Identifiants chimiques
Microplate Reader
USB Type B, USB Type A
Endpoint, Kinetic, Well-area Scanning
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
CE and TUV marked. RoHS compliant.
100/240 V
8 kg
Photodiode
Tungsten Halogen Lamp
Linear (except NB configurations)
10 nm
Measuring absorbance in microplate-based assays
18.5 lb.
Spécifications
Microplate Reader
USB Type B, USB Type A
Endpoint, Kinetic, Well-area Scanning
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
CE and TUV marked. RoHS compliant.
Measuring absorbance in microplate-based assays
800TSNB
400 to 750 nm
8 kg
Photodiode
Tungsten Halogen Lamp
Linear (except NB configurations)
10 nm
405, 450, 490, 630 nm
6- to 384-well Terasaki Plates
100/240 V
18.5 lb.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS