Learn More
Hamilton™ 800 Microliter™ Syringes, RN Termination
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
$249.57 - $1,594.49
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
For Use With (Application) Ideal for dispensing liquid to achieve the highest level of accuracy and precision possible, recommended for septum penetration
Length (English) 2 in.
Length (English) Needle 2 in.
View More Specs
Products 7
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Price Quantity & Availability  
14815269
View Documents Promotion Details
Hamilton™
HAM84890
10, 25, 50, 100, 250 μL 22s and 26 G
Each for $1,594.49
Only null left
14815205
View Documents Promotion Details
Hamilton™
HAM84855
25 μL 22s G
Each for $284.42
Only null left
14815227
missing translation for 'viewDocuments' Promotion Details
Hamilton™
HAM84859
100 μL 22s G
Each for $332.13
Only null left
1482480C
missing translation for 'viewDocuments' Promotion Details
Hamilton™
HAM84853
10 μL 26s G
Each for $249.57
Only null left
14815238
missing translation for 'viewDocuments' Promotion Details
Hamilton™
HAM84861
250 μL 22s G
Each for $357.83
Only null left
14815203
missing translation for 'viewDocuments' Promotion Details
Hamilton™
HAM84851
5 μL 26s G
Each for $282.58
Only null left
14815216
missing translation for 'viewDocuments' Promotion Details
Hamilton™
HAM84857
50 μL 22s G
Each for $310.11
Only null left
Description
Description
  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Specifications
Specifications
Autoclavable
Ideal for dispensing liquid to achieve the highest level of accuracy and precision possible, recommended for septum penetration
2 in.
2000 psi
2
Ea.
Removable
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
2 in.
51 mm
50°F
10°C
Sharp, Bevel, Curved, Non-coring