Learn More
Hamilton™ 800 Microliter™ Syringes, RN Termination
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
Supplier:  Hamilton™ HAM84853
 View more versions of this product
Catalog No. 1482480C
Only null left
Description
Description
  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Specifications
Chemical Identifiers
Autoclavable
Ideal for dispensing liquid to achieve the highest level of accuracy and precision possible, recommended for septum penetration
2 in.
2000 psi
2
Ea.
Removable
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
2 in.
51 mm
50°F
10°C
Sharp, Bevel, Curved, Non-coring
Specifications
Autoclavable
26s G
0.005 in.
0.0187 in.
0.48 mm
0.47 mm
Removable blue syringe holder
2 in.
Microliter Syringe
239°F
50°F
2
Ea.
Sharp, Bevel, Curved, Non-coring
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
0.01 in.
0.13 mm
0.26 in.
6.6 mm
Ideal for dispensing liquid to achieve the highest level of accuracy and precision possible, recommended for septum penetration
2 in.
51 mm
2000 psi
115°C
810
10°C
10 μL
Removable
Product Suggestions
Product Suggestions