Learn More
Eisco™ 8 Piece Tuning Fork Sets with Case
Catalog No. S27732
Change view
Click to view available options
Material:
Aluminum Tuning Fork
Steel Tuning Fork
Includes:
Plastic Case
Wooden Case
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Includes
S27732 Aluminum Tuning Fork Wooden Case
S27733 Steel Tuning Fork Plastic Case
S27731 Steel Tuning Fork Wooden Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S27732 Supplier Eisco™ Supplier No. PH0738B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
8 piece steel tuning forks with Scientific Pitch, C4 = 256Hz are designed for sound wave lessons and exploration
  • Supplied with either a plastic, or wooden case for long term storage
  • Tuning forks are nickel plated with plain shanks
  • Frequency is marked on each tuning fork
  • Includes 1 of each: C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512)
Specifications
Product Type Tuning Fork Set, 8 Piece
Description Scientific Pitch (C4 = 256 Hz)
Material Aluminum Tuning Fork
For Use With (Application) Sound Wave Lessons and Exploration
Includes Wooden Case