missing translation for 'onlineSavingsMsg'
Learn More

8-Pentadecanone 99.0+%, TCI America™

Catalog No. D15455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15455G 5 g
D154525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15455G Supplier TCI America Supplier No. D15455G

Chemical Identifiers

CAS 818-23-5
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.40
MDL Number MFCD00009555
InChI Key PQYGSSYFJIJDFK-UHFFFAOYSA-N
Synonym 8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
PubChem CID 13162
IUPAC Name pentadecan-8-one
SMILES CCCCCCCC(=O)CCCCCCC

Specifications

Melting Point 43°C
Color White
Boiling Point 140°C
Quantity 5 g
Formula Weight 226.40
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Pentadecanone
RTECSNumber RZ2000000
TSCA No