missing translation for 'onlineSavingsMsg'
Learn More

8-Oxoadénosine 98,0+%, TCI America™

Numéro de catalogue. o0401200mg
Modifier l'affichage
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
O0401200MG 200 mg
O04011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. O0401200MG Fournisseur TCI America Code fournisseur O0401200MG

Identifiants chimiques

CAS 29851-57-8
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.244
MDL Number MFCD15145291
InChI Key UEHOMUNTZPIBIL-DTUHVUQASA-N
Synonym 7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine
PubChem CID 91972088
IUPAC Name 6-amino-9-[(2R,3S,5R)-3,4-dihydroxy-5-(hydroxyméthyl)oxolan-2-yl]-7H-purine-8-one
SMILES C1=NC2=C(C(=N1)N)NC(=O)N2C3C(C(C(O3)CO)O)O

Spécifications

Color Blanc-jaune
Quantity 200 mg
Formula Weight 283.24
Percent Purity ≥98.0% (HPLC)
Physical Form Poudre cristalline
Chemical Name or Material 8-Oxoadenosine
TSCA No
Recommended Storage Réfrigérateur