missing translation for 'onlineSavingsMsg'
Learn More

8-Octanolactam, 98%

Catalog No. AAA1862122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1862122 100 g
AAA1862106 5 g
AAA1862114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1862122 Supplier Thermo Scientific Chemicals Supplier No. A1862122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 935-30-8 | C8H15NO | 141.214 g/mol

2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

Solubility
Soluble in methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 935-30-8
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00003273
InChI Key YDLSUFFXJYEVHW-UHFFFAOYSA-N
Synonym 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo
PubChem CID 13632
IUPAC Name azonan-2-one
SMILES C1CCCC(=O)NCCC1

Specifications

Melting Point 72°C to 75°C
Quantity 100 g
Beilstein 109775
Solubility Information Soluble in methanol (almost transparency).
Formula Weight 141.21
Percent Purity 98%
Chemical Name or Material 8-Octanolactam
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 213-300-3
RTECSNumber CO1757000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only