missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Methyl-6-nonenoic acid, predominantly trans, ≤15.0% (cis-8-Methyl-6-nonenoic acid); ≥80% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 59320-77-3 |
| Physical Form | Neat |
| Linear Formula | C10H18O2 |
| MDL Number | MFCD00203855 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 170.25 |
| Percent Purity | ≤15.0% (cis to 8 to Methyl to 6 to nonenoic acid); ≥80% (GC) |
| Grade | Analytical Standard |