missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyquinoline 98.0+%, TCI America™

Catalog No. M27181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27181G Supplier TCI America Supplier No. M27181G

Chemical Identifiers

CAS 938-33-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00957068
InChI Key ZLKGGEBOALGXJZ-UHFFFAOYSA-N
Synonym quinoline, 8-methoxy, methyl 8-quinolyl ether, ccris 7821, unii-4y56u1unuj, 4y56u1unuj, 8-methoxyquinoline, 8-methoxy quinoline, 8-methoxy-quinoline, acmc-209rns, ksc268q2n
PubChem CID 70310
IUPAC Name 8-methoxyquinoline
SMILES COC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 282°C
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Methoxyquinoline
TSCA No
Recommended Storage Refrigerator