missing translation for 'onlineSavingsMsg'
Learn More

8-Mercaptomenthone 97.0+%, TCI America™

Catalog No. M20235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20235G Supplier TCI America Supplier No. M20235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38462-22-5
Molecular Formula C10H18OS
Molecular Weight (g/mol) 186.313
MDL Number MFCD00012393
InChI Key RVOKNSFEAOYULQ-UHFFFAOYSA-N
Synonym p-mentha-8-thiol-3-one, 8-mercaptomenthone, 8-mercapto-p-menthan-3-one, thiomenthone, 8-mercapto-p-menthane-3-one, cyclohexanone, 2-1-mercapto-1-methylethyl-5-methyl, 2-1-mercapto-1-methylethyl-5-methylcyclohexanone, buchu mercaptan, 5-methyl-2-2-sulfanylpropan-2-yl cyclohexan-1-one, 2-1-mercapto-1-methylethyl-5-methylcyclohexan-1-one
PubChem CID 61982
IUPAC Name 5-methyl-2-(2-sulfanylpropan-2-yl)cyclohexan-1-one
SMILES CC1CCC(C(=O)C1)C(C)(C)S

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 186.31
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 8-Mercaptomenthone