missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline Sulfate, 98%3, Spectrum™ Chemical

Catalog No. 18608298
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608298 50 kg Amber Glass Bottle
18608293 25 g Amber Glass Bottle
18608294 100 g Amber Glass Bottle
18608295 500 g Amber Glass Bottle
18608296 2.5 kg Amber Glass Bottle
18608297 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608298 Supplier Spectrum Chemical Mfg Cor Supplier No. H109650KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134-31-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 50 kg
Molecular Weight (g/mol) 388.4
Percent Purity 0.983
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 8-Hydroxyquinoline Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.