missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-5-nitroquinoline 98.0+%, TCI America™

Catalog No. H080525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H080525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H080525G Supplier TCI America Supplier No. H080525G

Chemical Identifiers

CAS 4008-48-4
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD00006791
InChI Key RJIWZDNTCBHXAL-UHFFFAOYSA-N
Synonym nitroxoline, 5-nitroquinolin-8-ol, 8-hydroxy-5-nitroquinoline, 5-nitro-8-hydroxyquinoline, 5-nitro-8-quinolinol, nitroxolin, noxibiol, 5-nitrox, 5-nitroxine, 5-nitroks
PubChem CID 19910
ChEBI CHEBI:67121
SMILES C1=CC2=C(C=CC(=C2N=C1)O)[N+](=O)[O-]

Specifications

Melting Point 180°C
Quantity 25 g
Formula Weight 190.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxy-5-nitroquinoline
RTECSNumber VC8225000
TSCA No