missing translation for 'onlineSavingsMsg'
Learn More

8-Chloroquinoline 98.0+%, TCI America™

Catalog No. C02845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02845G 5 g
C028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02845G Supplier TCI America Supplier No. C02845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-33-6
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00047618
InChI Key RUSMDMDNFUYZTM-UHFFFAOYSA-N
Synonym quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
PubChem CID 69139
ChEBI CHEBI:48985
IUPAC Name 8-chloroquinoline
SMILES ClC1=C2N=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 288°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Chloroquinoline