missing translation for 'onlineSavingsMsg'
Learn More

8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine, 97%, Thermo Scientific™

Numéro de catalogue. p-2703556
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CD08167CB 250mg
CD08167DA 1g
CD08167EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CD08167CB Fournisseur Thermo Scientific Chemicals Code fournisseur CD08167CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 131728-94-4
Formule moléculaire C9H8ClFO2
Poids moléculaire (g/mol) 202.61
Numéro MDL MFCD00052603
Clé InChI FMONGDHUPLQOCP-UHFFFAOYSA-N
Synonyme 8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
PubChem CID 2780137
Nom de l’IUPAC 8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine
SOURIRES FC1=CC(CCl)=C2OCOCC2=C1

Spécifications

Nom ou substance chimique 8-(Chloromethyl)-6-fluoro-4H-1, 3-benzodioxine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 202.61
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.