missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH3509403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3509403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3509403 Supplier Thermo Scientific Chemicals Supplier No. H3509403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 957187-27-8 | C7H4BrClN2 | 231.48 g/mol

8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 957187-27-8
Molecular Formula C7H4BrClN2
Molecular Weight (g/mol) 231.48
MDL Number MFCD12828062
InChI Key LMPPBTNPACXNDI-UHFFFAOYSA-N
Synonym 8-bromo-6-chloroimidazo 1,2-a pyridine, 8-bromo-6-chloro-imidazo 1,2-a pyridine, 6-chloro-8-bromoimidazo 1,2-a pyridine
PubChem CID 26370038
IUPAC Name 8-bromo-6-chloroimidazo[1,2-a]pyridine
SMILES ClC1=CN2C=CN=C2C(Br)=C1

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 231.48
Percent Purity 95%
Chemical Name or Material 8-Bromo-6-chloroimidazo[1,2-a]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only