missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-2'-deoxyguanosine, 99%

Catalog No. AAJ6472303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6472303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6472303 Supplier Thermo Scientific Chemicals Supplier No. J6472303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13389-03-2 | C10H12BrN5O4 | 346.14 g/mol

8-Bromo-2'-deoxyguanosine is useful as the starting structure for 2'-deoxyguanosine nucleotides, which are modified in position 8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-2′-deoxyguanosine is useful as the starting structure for 2′-deoxyguanosine nucleotides, which are modified in position 8.

Solubility
Soluble in dimethyl sulfoxide, warm water and dimethyl formam

Chemical Identifiers

CAS 13389-03-2
Molecular Formula C10H12BrN5O4
Molecular Weight (g/mol) 346.14
MDL Number MFCD01630971
InChI Key MKDXZFVCXWXGBQ-QXZWWJJYNA-N
Synonym 8-bromo-2'-deoxyguanosine, 8-bromo-2'-dg, 2-amino-8-bromo-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 8-br-2'-dg, guanosine, 8-bromo-2'-deoxy, 8-bromodeoxyguanosine, epitope id:151524, 8-bromo-2?-deoxyguanosine, guanosine,8-bromo-2'-deoxy, 8-bromo-2-deoxyguanosine
PubChem CID 10947964
ChEBI CHEBI:61072
IUPAC Name 2-amino-8-bromo-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=C(Br)N2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

Melting Point >250°C (decomposition)
Color White
Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide,warm water and dimethyl formamide.
Formula Weight 346.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 8-Bromo-2'-deoxyguanosine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only