missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromo-1-octanol 90,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Quantity | 5 g |
| Formula Weight | 209.13 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 8-Bromo-1-octanol |
Sécurité et manipulation
| TSCA | Yes |
| Recommended Storage | Réfrigérateur |