missing translation for 'onlineSavingsMsg'
Learn More

8-Azahypoxanthine, 98%

Catalog No. AC202590050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC202590050 5 g
AC202590010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC202590050 Supplier Thermo Scientific Chemicals Supplier No. 202590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2683-90-1 | C4H3N5O | 137.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2683-90-1
Molecular Formula C4H3N5O
Molecular Weight (g/mol) 137.10
MDL Number MFCD00005804
InChI Key OEEYCNOOAHGFHL-UHFFFAOYSA-N
Synonym 8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine
PubChem CID 75895
IUPAC Name 2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
SMILES O=C1N=CN=C2NNN=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 26, IV, 4130
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution at ph >9
Formula Weight 137.1
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 8-Azahypoxanthine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 220-244-3
TSCA TSCA

RUO – Research Use Only