missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Azaadenine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 320°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 136.12 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 8-Azaadenine |
Safety and Handling
| RTECSNumber | XZ5680000 |
| TSCA | Yes |